Key Documents
AXYRESSW96HPSI
AxyVoir reservoir
96 bottom, high profile single well, individually wrapped, 25 ea of, sterile
About This Item
UNSPSC Code:
41121522
NACRES:
NB.22
sterility
sterile
manufacturer/tradename
Axygen Inc RES-SW96-HP-SI
Corning
packaging
25 ea of
General description
  • Axygen® reagent reservoir is a multiple-well high-profile reservoir manufactured using chemically resistant polypropylene.
  • Ideal for both manual and automated applications
  • Designed with standard ANSI/SLAS footprint dimensions for microplates.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.