Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV47712

Sigma-Aldrich

Anti-BATF2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Basic leucine zipper transcription factor, ATF-like 2, Anti-MGC20410

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

20 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BATF2(116071)

General description

BATF2 is a leucine zipper protein that may function as a transcription factor. Decreased BATF2 expression has been linked to the poor prognosis of oral tongue squamous cell and hepatocellular carcinomas.
Rabbit anti-rabbit BATF2 antibody recognizes human and canine BATF2.

Immunogen

Synthetic peptide directed towards the middle region of human BATF2

Application

Rabbit anti-rabbit BATF2 antibody is suitable for western blot applications at a concentration of 0.25μg/ml.

Sequence

Synthetic peptide located within the following region: AQTAPPQPLELEHPTRGKLGSSPDNPSSALGLARLQSREHKPALSAATWQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haojie Wen et al.
Oncology reports, 31(1), 169-174 (2013-11-21)
BATF2, also called SARI, is associated with several cancer types, and loss of BATF2 expression is frequently detected in aggressive and metastatic cancers. The expression of BATF2 was previously shown to slow the growth rate of malignant tumor cells injected
Haiqing Ma et al.
International journal of cancer, 128(4), 771-777 (2010-05-18)
Human BATF2, a basic leucine zipper protein, was recently detected in several normal immortalized cell lines but not in transformed cell lines. In addition, the expression of BATF2 also slowed the growth rate of malignant tumor cells injected into athymic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service