Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV32589

Sigma-Aldrich

Anti-CHEK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CHK1 checkpoint homolog (S. pombe)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

rat, human, rabbit, mouse, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHEK1(1111)

General description

CHEK1 is a serine/threonine kinase that functions as a sensor for the stress response pathway in cells. It is activated by ATR kinase phosphorylation in response to DNA damage, and can subsequently modulate p53 phosphorylation. CHEK1 activity is inhibited by BCL6 in B cells.
Rabbit Anti-CHEK1 antibody recognizes zebrafish, bovine, chicken, human, mouse, rat, and canine CHEK1.

Immunogen

Synthetic peptide directed towards the middle region of human CHEK1

Application

Rabbit Anti-CHEK1 antibody can be used for western blot applications at a concentration of 0.5μg/ml. It can also be used for immunohistochemistry at 4-8μg/ml.

Biochem/physiol Actions

CHEK1 is required during normal S phase to avoid aberrantly increased initiation of DNA replication, thereby protecting against DNA breakage. Its expression is dispensable for somatic cell death and critical for sustaining G2 DNA damage checkpoint.

Sequence

Synthetic peptide located within the following region: WSCGIVLTAMLAGELPWDQPSDSCQEYSDWKEKKTYLNPWKKIDSAPLAL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Li et al.
Nature communications, 12(1), 3845-3845 (2021-06-24)
Atr is a serine/threonine kinase, known to sense single-stranded DNA breaks and activate the DNA damage checkpoint by phosphorylating Chek1, which inhibits Cdc25, causing cell cycle arrest. This pathway has not been implicated in neuroregeneration. We show that in Drosophila
Hala Gali-Muhtasib et al.
Cancer research, 68(14), 5609-5618 (2008-07-18)
There are few reports describing the role of p53-dependent gene repression in apoptotic cell death. To identify such apoptosis-associated p53 target genes, we used the pro-oxidant plant-derived drug thymoquinone and compared p53+/+ and p53-/- colon cancer cells HCT116. The p53
Stella M Ranuncolo et al.
Blood cells, molecules & diseases, 41(1), 95-99 (2008-03-19)
BCL6 is a transcriptional repressor protein that is expressed in a developmentally regulated fashion during B-cell maturation. Specifically, BCL6 is required for formation of germinal centers in response to T-cell dependent antigen activation. Germinal center B-cells feature the ability to
Hanna Engqvist et al.
Frontiers in oncology, 10, 162-162 (2020-03-07)
Early-stage (I and II) ovarian carcinoma patients generally have good prognosis. Yet, some patients die earlier than expected. Thus, it is important to stratify early-stage patients into risk groups to identify those in need of more aggressive treatment regimens. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service