Skip to Content
MilliporeSigma

Skip To

A9101

Acetylcholine chloride

≥99% (TLC), powder or crystals

Synonym(s):

ACh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 vials

Available to ship TODAYfromMILWAUKEE

$358.00

About This Item

Linear Formula:
(CH3)3N+CH2CH2OCOCH3Cl-
CAS Number:
Molecular Weight:
181.66
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
200-468-8
MDL number:
Beilstein/REAXYS Number:
3571875
eCl@ss:
32190102

$358.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetylcholine chloride, pkg of 150 mg (per vial)

assay

≥99% (TLC)

Quality Segment

form

powder or crystals

packaging

pkg of 150 mg (per vial)

mp

146-150 °C (lit.)

solubility

H2O: soluble

storage temp.

room temp

SMILES string

[Cl-].CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

JUGOREOARAHOCO-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

General description

Acetylcholine is an important neurotransmitter that plays a critical role in multiple neurological systems. Crucial for neurobiology studies, acetylcholine aids researchers in understanding synaptic transmission. In other forms of research, acetylcholine has an effect and influence on tumor neurobiology for cancer R&D, as well as a variety of other cognitive processes.

Application

For use as a substrate to determine acetylcholinesterase activity. Acetylcholine chloride has been used:
  • for standard curve preparation in hestrin assay
  • in Protein expression, radioactive ligand binding assay and electrophysiological studies of Lymnaea stagnalis acetylcholine binding protein (Ls-AChBPs)
  • as a supplement in extracellular solutions for treating the voltage clamped cells in electrophysiology studies

Biochem/physiol Actions

Acetyl choline (ACh) is an essential neurotransmitter in both central and peripheral nervous system. ACh binds to nicotinic acetylcholine receptors (nAChRs) and causes fast excitatory neurotransmission.
Acetylcholine chloride, injected at 20 mg/kg body weight, reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis . The cholinergic anti-inflammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.
Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Preparation Note

Reconstituted solution is stable for 2 weeks when stored at 2-8 °C

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR15461008501A6625
assay

≥99% (TLC)

assay

-

assay

-

assay

≥99% (TLC)

solubility

H2O: soluble

solubility

-

solubility

-

solubility

H2O: 100 mg/mL

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

200

form

powder or crystals

form

-

form

-

form

-

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

mp

146-150 °C (lit.)

mp

146-150 °C (lit.)

mp

146-150 °C (lit.)

mp

146-150 °C (lit.)


Still not finding the right product?

Explore all of our products under Acetylcholine chloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters