Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0233

Sigma-Aldrich

AS605240

≥98% (HPLC)

Synonym(s):

5-(6-Quinoxalinylmethylene)-2,4-thiazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7N3O2S
CAS Number:
Molecular Weight:
257.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, faint red to orange to dark brown

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1NC(=O)C(\S1)=C\c2ccc3nccnc3c2

InChI

1S/C12H7N3O2S/c16-11-10(18-12(17)15-11)6-7-1-2-8-9(5-7)14-4-3-13-8/h1-6H,(H,15,16,17)/b10-6-

InChI key

SQWZFLMPDUSYGV-POHAHGRESA-N

Application

AS605240 has been used:
  • as a phosphatidylinositol 3-kinase (PI3K) inhibitor to study its effect on Rab5-positive early endosome (EE) vesicles.
  • as a (PI3K) inhibitor to treat splenocytes for graft-versus-host disease (GVHD) induction.
  • as a class 1A PI3K inhibitor to treat CD4+ T cells.

Biochem/physiol Actions

AS605240 inhibits human recombinant phosphatidylinositol 3-kinase (PI3K) γ, α, β, and δ by competing with adenosine triphosphate (ATP). It plays a role in protecting injury following ischemic stroke. AS605240 also inhibits the activation of astrocytes under the influence of Interleukin-6 (IL-6) and its soluble receptor (sIL-6R). It reduces collagen deposition and prevents lung inflammation. AS605240 also inhibits the increase of tumor necrosis factor-α (TNF-α) and interleukin-1β (IL-1β) expression instigated by bleomycin. It is effective in preventing pulmonary fibrosis by inhibiting the transforming growth factor β1 (TGF-β1) and T lymphocytes infiltration into lungs.
AS605240 is a potent and selective PI3Kγ inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphoinositide Kinases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyi Yu et al.
Scientific reports, 8(1), 10988-10988 (2018-07-22)
Chronic kidney disease (CKD) poses a formidable challenge for public healthcare worldwide as vast majority of patients with CKD are also at risk of accelerated cardiovascular disease and death. Renal fibrosis is the common manifestation of CKD that usually leads
Irene Filippi et al.
Journal of cellular physiology, 229(12), 2067-2076 (2014-05-14)
Hypoxia represents an inadequate oxygen supply to tissues, which can modulate cell functions, primarily through the hypoxia-inducible transcription factor HIF-1α. Dendritic cells (DC) are professional antigen-presenting cells and their migration maybe affected by hypoxia, since the local microenvironment in lymphoid
Cheng-Chang Chen et al.
Cell chemical biology, 24(7), 907-916 (2017-07-22)
To resolve the subcellular distribution of endolysosomal ion channels, we have established a novel experimental approach to selectively patch clamp Rab5 positive early endosomes (EE) versus Rab7/LAMP1-positive late endosomes/lysosomes (LE/LY). To functionally characterize ion channels in endolysosomal membranes with the patch-clamp
Sen Shang et al.
Neuroscience, 415, 107-120 (2019-06-14)
The intense and prolonged inflammatory response after ischemic stroke significantly contributes to the secondary neural injury. PI3Kγ, which is involved in the regulation of vascular permeability, chemotactic leukocyte migration and microglia activation, is a key target for intervention in the
Xin Wei et al.
Biochemical and biophysical research communications, 397(2), 311-317 (2010-06-01)
Phosphoinositide 3-kinase-gamma (PI3Kgamma) has been identified to play the critical roles in inflammatory cells activation and recruitment in multiply inflammatory diseases and it promised to be a prospective target for relevant inflammatory diseases therapy. AS605240, a selective PI3Kgamma inhibitor, has

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service