It is not tested for this application. This product is only tested on human tissue for Immunohistochemistry. It may be possible for it to work on other species but that is not the listed application.???????
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Please contact Customer Service for Availability | $550.00 |
About This Item
NACRES:
SB.01
UNSPSC Code:
12352203
Technique(s):
immunohistochemistry: suitable
Storage temp.:
2-8°C
$550.00
Please contact Customer Service for Availability
description
For In Vitro Diagnostic Use in Select Regions
Quality Segment
form
aqueous solution
manufacturer/tradename
Cell Marque®
packaging
kit of (7 mL each of predilute amplifier and polymer detector (954D-10)), kit of (containing 100 mL each of predilute amplifier and polymer detector (954D-30)), kit of (containing 1000 mL each of predilute amplifier and polymer detector (954D-40)), kit of (containing 15 mL each of predilute amplifier and polymer detector (954D-50)), kit of (containing 50 mL each of predilute amplifier and polymer detector (954D-20))
technique(s)
immunohistochemistry: suitable
storage temp.
2-8°C
General description
The HiDef Detection Polymer System is a visualization system that is ready-to-use in immunohistochemical protocols. This two-step system uses an indirect method resulting in an antibody-enzyme complex that universally detects mouse and rabbit primary antibodies. The resulting chromogenic reaction can be visualized by HRP-compatible chromogens using light microscopy. It is biotin-free and eliminates non-specific staining that could result from endogenous biotin. This visualization system consists of two detection reagents and is based on the sequential application of HiDef Detection Amplifier (Mouse and Rabbit) followed by HiDef Detection HRP Polymer Detector, amplifying the detection of low expressing antigens. This detection system is compatible with both manual and open automated staining platforms.
Physical form
Ready to use reagent
Preparation Note
Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.
Note: This requires a keycode which can be found on your packaging or product label.
Analysis Note
United States - IVD
Canada - IVD
European Union - IVD
Japan - RUO
Canada - IVD
European Union - IVD
Japan - RUO
Other Notes
For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com
Legal Information
Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hello, my question is the following, is the reagent II (HRP HiDef Detection™ HRP Polymer Detector) compatible with the use of secondary antibodies from other species? In our laboratory we not only use rabbit-mouse, but some are also goat or rat secondary
1 answer-
Helpful?
-

