Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420M-1

Sigma-Aldrich

p120 Catenin (MRQ-5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (420M-14)
vial of 0.5 mL concentrate (420M-15)
bottle of 1.0 mL predilute (420M-17)
vial of 1.0 mL concentrate (420M-16)
bottle of 7.0 mL predilute (420M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG1

control

lobular breast carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... CTNND1(1500)

General description

p120 catenin is encoded on chromosome 11q11. Alpha-catenin and beta-catenin bind to the intracellular domain of E-cadherin while p120 catenin binds E-cadherin at a juxta-membrane site. The complex stabilizes tight junctions. In the cell, p120 catenin localized to the E-cadherin/catenins cell adhesion complex, directly associates with cytoplasmic C-terminus of E-cadherin and may similarly interact with other cadherins. A deficiency of E-cadherin results in the intracytoplasmic accumulation of p120 catenin. Lobular carcinoma of the breast shows intracytoplasmic accumulation of p120 catenin while ductal carcinoma shows reduced membrane p120 catenin without cytoplasmic accumulation. In gastric and colonic carcinoma, strong cytoplasmic p120 catenin is associated with discohesive infiltrative morphology.

Quality


IVD

IVD

IVD

RUO

Linkage

p120 Catenin Positive Control Slides, Product No. 420S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David I Bellovin et al.
Cancer research, 65(23), 10938-10945 (2005-12-03)
We examined the expression and localization of p120 catenin (p120ctn) as a consequence of the epithelial to mesenchymal transition (EMT) of highly differentiated colon carcinoma cells (LIM1863 cells). This unique line grows in suspension as spheroids and undergoes an EMT
A U Jawhari et al.
The Journal of pathology, 189(2), 180-185 (1999-11-05)
p120(ctn) is a substrate of the tyrosine kinase pp60 src. Tyrosine kinases such as src localize to the adherens junctions and phosphorylate junctional proteins in both normal and transformed cells.(1) p120(ctn) forms a complex with E-cadherin at the adherens junction
David J Dabbs et al.
The American journal of surgical pathology, 31(3), 427-437 (2007-02-28)
The distinction between lobular and ductal lesions of the breast is important in several circumstances. Diagnostic reproducibility of lobular versus ductal lesions, based on histology alone, is less than optimal. The proper distinction between atypical lobular hyperplasia, lobular carcinoma in
A B Reynolds et al.
Oncogene, 7(12), 2439-2445 (1992-12-01)
A novel protein tyrosine kinase (PTK) substrate, p120, has been previously implicated in ligand-induced signaling through the epidermal growth factor, platelet-derived growth factor and colony-stimulating factor 1 receptors, and in cell transformation by p60v-src. We have isolated a near full-length
David Sarrió et al.
Oncogene, 23(19), 3272-3283 (2004-04-13)
Accumulating evidences indicate that p120 catenin, a member of the E-cadherin (E-CD)/catenin adhesion complex, plays a role in tumor invasion. To establish the expression pattern of p120 in breast cancer, we analysed 326 breast tissue biopsies by tissue microarray. Most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service