Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24700C

SAFC

EX-CELL® CD Hydrolysate Fusion

Animal-component free, powder, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

Quality Level

description

for research or for further manufacturing use

form

powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EX-CELL® CD Hydrolysate Fusion is a synthetic option to undefined hydrolysates used in cell culture processes, including both Batch and Fed-Batch modes. It is formulated to have minimal pH and osmolality impact upon addition. EX-CELL CD Hydrolysate Fusion is optimized for Chinese Hamster Ovary (CHO) cell lines but has also been shown to work well with other cell types (e.g. NS0 and Sp2/0). EX-CELL CD Hydrolysate Fusion is comprised only of animal-free, chemically defined components that were identified to be in hydrolysates (but synthetically produced) or are commonly found in cell culture media. In order to have broad applications, EX-CELL CD Hydrolysate Fusion was based on data generated from four hydrolysate types (soy, wheat gluten, yeast extract and meat).

Application

EX-CELL® CD Hydrolysate Fusion has been used to determine its effect on monoclonal antibody productivity, purification and quality in Chinese hamster ovary (CHO) cells.
EX-CELL® CD Hydrolysate Fusion is a synthetic option to undefined hydrolysates used in cell culture processes, including both Batch and Fed-Batch modes. It is formulated to have minimal pH and osmolality impact upon addition. EX-CELL CD Hydrolysate Fusion is optimized for Chinese Hamster Ovary (CHO) cell lines but has also been shown to work well with other cell types (e.g. NS0 and Sp2/0). EX-CELL CD Hydrolysate Fusion is comprised only of animal-free, chemically defined components that were identified to be in hydrolysates (but synthetically produced) or are commonly found in cell culture media. In order to have broad applications, EX-CELL CD Hydrolysate Fusion was based on data generated from four hydrolysate types (soy, wheat gluten, yeast extract and meat).

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of hydrolysates on monoclonal antibody productivity, purification and quality in Chinese hamster ovary cells.
Ho S C, et al.
Journal of Bioscience and Bioengineering, 122(4), 499-506 (2016)

Articles

Chemically defined media supplements offer many benefits including decreased lot-to-lot variation, decreased risks, and improved glycosylation profiles.

Why optimize your upstream bioprocess media and feed? This article discusses the why and how of optimizing your media and feed to maximize cell viability and productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service