Skip to Content
MilliporeSigma
All Photos(1)

Documents

231M-1

Sigma-Aldrich

Calponin (CALP) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

CALP, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (231M-14)
vial of 0.5 mL concentrate (231M-15)
bottle of 1.0 mL predilute (231M-17)
vial of 1.0 mL concentrate (231M-16)
bottle of 7.0 mL predilute (231M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

appendix

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Calponin is a 34 kD polypeptide that interacts with actin, tropomyosin, and calmodulin. It is involved in smooth muscle contraction mechanism and is restricted exclusively to smooth muscle tissue. Anti-calponin has been found to be useful in differentiating benign sclerosing lesions of the breast from carcinoma. Calponin positivity has also been noted in malignant myoepithelioma and pleomorphic adenoma of salivary gland origin, as well as angiomatoid malignant fibrous histiocytoma.

Quality


IVD

IVD

IVD

RUO

Linkage

Calponin Positive Control Slides, Product No. 231S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibodies to novel myoepithelium-associated proteins distinguish be-nign lesions and carcinoma in situ from invasive carcinoma of the breast
Wang NP, Wan BC, et al.
Appl. Immunohistochem., 5(3), 141-151 (1997)
A T Savera et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 10(11), 1093-1100 (1997-12-05)
Myoepithelial cells of salivary glands have a complex cytoskeletal immunophenotype. To elaborate the smooth muscle phenotype of salivary gland myoepithelium and to assess its contribution to the histogenesis of pleomorphic adenomas, we evaluated the immunohistochemical expression of three novel monoclonal
J C Fanburg-Smith et al.
Human pathology, 30(11), 1336-1343 (1999-11-26)
Angiomatoid "malignant" fibrous histiocytoma (AMFH) has been considered to be a low-grade sarcoma of childhood, and, with its fibrous pseudocapsule, angiomatoid change, dense lymphoplasmacytic response, and proliferation of spindled or round cells, has been classified as a fibrohistiocytic neoplasm. We
Jason L Hornick et al.
The American journal of surgical pathology, 27(9), 1183-1196 (2003-09-10)
Myoepitheliomas and mixed tumors were only recently recognized to occur primarily in soft tissue, and only small case numbers have been described. To characterize these tumors further and to evaluate prognostic parameters, 101 myoepithelial tumors of soft tissue were retrieved
T Nagao et al.
Cancer, 83(7), 1292-1299 (1998-10-08)
Malignant myoepithelioma (MME) of the salivary gland, also known as myoepithelial carcinoma, is rare and its biologic behavior has not been clarified fully. Ten cases of MME were analyzed for their clinicopathologic features and immunohistochemical characteristics, focusing on prognostic factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service