Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

137259

SAFC

PIPES disodium salt

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

PIPES disodium salt, 1,4-Piperazinediethanesulfonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2Na2O6S2
CAS Number:
Molecular Weight:
346.33
Beilstein:
9453872
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin and microbial; tested

pH

6.1-7.5

useful pH range

6.1-7.5

pKa (25 °C)

6.8

pKa 

6.8

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2Na/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

GMHSTJRPSVFLMT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

PIPES Disodium is the sodium salt form of PIPES and is a biological buffer often referred to as a “Good′s” buffer. The pKa of PIPES is 6.8 which makes it ideal for biological applications at physiological conditions. PIPES Disodium is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.PIPES Disodium can be used in a wide array of biological applications including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service