Skip to Content
MilliporeSigma
All Photos(3)

Documents

31454

Sigma-Aldrich

Sodium sulfite

puriss. p.a., reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

grade

puriss. p.a.

Assay

98-100.0% (iodometric)

form

solid

impurities

≤0.001% heavy metals (as Pb)
≤0.02% thiosulfate (S2O3)

pH

9.0-10.5 (25 °C, 126 g/L)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

As: ≤0.2 mg/kg
Ca: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Pb: ≤5 mg/kg
Se: ≤10 mg/kg
Zn: ≤10 mg/kg

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfie is an inorganic sodium salt. Rapid-mixing method has been proposed to investigate kinetics of the reaction between oxygen and sodium sulfite.

Application

Sodium sulfite was used in the preparation of films based on wheat gluten.

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction kinetics of sodium sulfite oxidation by the rapid-mixing method.
Barron CH and O'Hern HA.
Chemical Engineering Science, 21(5), 397-404 (1966)
New plasticizers for wheat gluten films.
Irissin-Mangata J, et al.
European Polymer Journal, 37(8), 1533-1541 (2001)
Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Jingang Wang et al.
Chemosphere, 91(3), 351-357 (2012-12-26)
This paper presents a simple, easy and reliable method for determination of EDTA (ethylenediaminetetraacetic acid) in aqueous system. Using EDTA chelating with ferric irons, an excessive amount of Fe(3+) was added to EDTA solution, and Fe(3+) in excess was reduced
Fabian Leinisch et al.
Chemical research in toxicology, 24(12), 2217-2226 (2011-10-19)
Nitrone spin traps such as 5,5-dimethyl-1-pyrroline-N-oxide (DMPO) are commonly used for free radical detection. Though proven examples are rare, artifact formation must be considered. For example, the Forrester-Hepburn mechanism yields the same radical adduct as that formed by genuine radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service