Skip to Content
MilliporeSigma
All Photos(2)

Documents

270547

Sigma-Aldrich

N,N-Dimethylformamide

suitable for HPLC, ≥99.9%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

833 °F

purified by

glass distillation

expl. lim.

15.2 %

technique(s)

HPLC: suitable

impurities

≤0.030% water

evapn. residue

<0.0005%

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

λ

H2O reference

UV absorption

λ: 270 nm Amax: 1.00
λ: 275 nm Amax: 0.30
λ: 295 nm Amax: 0.10
λ: 310 nm Amax: 0.05
λ: 340-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and electrochemical characterization of aPEO-based polymer electrolytes.
Rodrigues LC, et al.
Journal of Solid State Electrochemistry, 16(4), 1623-1629 (2012)
Core-Shell Tubular Nanostructured Electrode of Hollow Carbon Nanofiber/Manganese Oxide for Electrochemical Capacitors.
Hong S, et al.
Electrochimica Acta, 141, 39-44 (2014)
W P Inskeep et al.
Plant physiology, 77(2), 483-485 (1985-02-01)
We found inconsistencies in the commonly used data for chlorophyll analysis in 80% acetone. Recently developed extinction coefficients for chlorophyll b in N,N-dimethylformamide (DMF) based on values from 80% acetone are low as a result of these inconsistencies. We determined
Kun Liu et al.
Nanoscale, 4(20), 6574-6580 (2012-09-15)
Studies on the self-assembly of metal nanoparticles (NPs) in the presence of ions are motivated by the biosensing applications of NP clusters and the capability to control the morphology of clusters of oppositely charged NPs. The effect of ions has
Leonard Blaschek et al.
Frontiers in plant science, 11, 109-109 (2020-03-21)
Lignin accumulates in the cell walls of specialized cell types to enable plants to stand upright and conduct water and minerals, withstand abiotic stresses, and defend themselves against pathogens. These functions depend on specific lignin concentrations and subunit composition in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service