Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

225673

Sigma-Aldrich

Silver sulfate

ACS reagent, 99%

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

99%

form

lumps and powder

reaction suitability

reagent type: catalyst
core: silver

impurities

≤0.02% insoluble matter and silver chloride
≤0.03% not HCl pptd.

mp

652 °C (lit.)

anion traces

nitrate (NO3-): ≤0.001%

cation traces

Fe: ≤0.001%

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Silver sulfate is an inorganic compound that can be used as a catalyst in various reactions such as halodecarboxylation of aryl aromatic acids, ortho-acrylation, and cycloaddition reaction to synthesize N-vinyl-substituted 1,2,3-triazoles.

Application

Silver sulfate can be used as:
  • A catalyst for the sulfonation of waste-based polystyrene into polystyrene sulfonic acid.
  • A chloride inhibitory agent in the determination of chemical oxygen demand (COD).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of the amount of catalyst on the degree of sulfonation in the sulfonation process on waste-based Styrofoam (polystyrene) into Poly-(Styrene Sulfonic Acid)
Manurung R, et al.
IOP Conference Series: Materials Science and Engineering, 1122, 012100-012100 (2021)
Rhodium-catalyzed ortho-acrylation of aryl ketone O-methyl oximes with cyclopropenones
Liu Y, et al.
Organic & Biomolecular Chemistry, 18 (2020)
Inexpensive NaX (X= I, Br, Cl) as a halogen donor in the practical Ag/Cu-mediated decarboxylative halogenation of aryl carboxylic acids under aerobic conditions
Fu Z, et al.
Organic & Biomolecular Chemistry, 16 (2018)
Phenol and Colour Removal from Wastewater Using Two Coal-Based Adsorptive Materials: Preliminary Laboratory Tests and Performances
ZAHARIA C
Bulletin of the Polytechnic Institute of jassy, Section Chemistry and Chemical Engineering, 61, 11-22 (2016)
Silver-mediated three-component cycloaddition reaction for direct synthesis of 1-N-vinyl-substituted 1, 2, 3-triazoles
Organic & Biomolecular Chemistry, 17, 4843-4849 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service