Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

223417

Sigma-Aldrich

Sodium peroxide

granular, +140 mesh particle size, reagent grade, 97%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.55

grade

reagent grade

Assay

97%

form

granular

reaction suitability

reagent type: oxidant

particle size

+140 mesh

pH

12.8 (20 °C, 100 g/L)

mp

460 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium peroxide (Sodium superoxide) has been synthesized by the matrix reaction of sodium atoms and oxygen molecules at high dilution in argon. Its IR spectral studies have been reported. It is a strong oxidizing agent.
Sodium peroxide is a strong oxidizing and bleaching agent mainly used in bleaching papers and textiles. It is an effective oxidizing flux for the decomposition of minerals and ores.

Application

Sodium peroxide may be used as an oxidant for:
conversion of Δ4-3-ketosteroids to Δ4-3,6-diones
oxidation of alduronic and glyculosonic acids
Sodium peroxide may be used for the isotope dilution inductively coupled plasma mass spectrometric quantification of Ru, Pd, Ir and Pt in various geological samples. It may be employed for the inductively coupled plasma mass spectrometric quantification of rare earth elements (such as Y, Th, Zr, Hf, Nb and Ta) in geological reference materials.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wiberg E and Wiberg N
Inorganic Chemistry, 502-502 (2001)
Oxidation of sodium salts of alduronic and glyculosonic acids by sodium peroxide.
Isbell HS, et al.
Carbohydrate Research, 36(2), 283-291 (1974)
Oxidation with sodium peroxide Direct introduction of a ? carbonyl group into a, ? unsaturated ketosteroids
Tetrahedron Letters, 22(51), 5127-5128 (1981)
Eagleson M.
Concise Encyclopedia Chemistry, 1080-1080 (1994)
Infrared spectra and bonding in the sodium superoxide and sodium peroxide molecules.
Andrews L.
The Journal of Physical Chemistry, 73(11), 3922-3928 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service