Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001600

Metformin impurity F

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH · HCl
CAS Number:
Molecular Weight:
81.54
Beilstein:
3589311
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metformin

form

solution

manufacturer/tradename

EDQM

mp

170-173 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

mixture

storage temp.

2-8°C

SMILES string

Cl[H].CNC

InChI

1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H

InChI key

IQDGSYLLQPDQDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metformin impurity F EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service