Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001559

Salvianolic acid B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S,3S)-4-[(1E)-3-[(1R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy]-3-oxo-1-propen-1-yl]-2-(3,4-dihydroxyphenyl)-2,3-dihydro-7-hydroxy-3-benzofurancarboxylic acid 3-[(1R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl] ester, Dan Shen Suan B, Danfensuan B, Lithospermic acid B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30O16
CAS Number:
Molecular Weight:
718.61
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salvianolic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c3O[C@@H]([C@@H](C(=O)O[C@H](Cc4ccc(O)c(O)c4)C(O)=O)c23)c5ccc(O)c(O)c5

InChI

1S/C36H30O16/c37-20-6-1-16(11-24(20)41)13-27(34(45)46)50-29(44)10-5-18-3-9-23(40)33-30(18)31(32(52-33)19-4-8-22(39)26(43)15-19)36(49)51-28(35(47)48)14-17-2-7-21(38)25(42)12-17/h1-12,15,27-28,31-32,37-43H,13-14H2,(H,45,46)(H,47,48)/b10-5+/t27-,28-,31+,32-/m1/s1

InChI key

SNKFFCBZYFGCQN-VWUOOIFGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salvianolic acid B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Hong Du et al.
World journal of gastroenterology, 20(44), 16674-16682 (2014-12-04)
To investigate the effects of Glytan on splanchnic hemodynamics and its reduction of portal pressure in portal hypertensive rats. Glytan (Ganluotong in Chinese), is composed of salvianolic acid B and diammonium glycyrrhizinate. Portal hypertension (PHT) was induced in the rats
Chunli Zheng et al.
Molecular diversity, 18(3), 621-635 (2014-05-06)
The term systems pharmacology describes a field of study that uses computational and experimental approaches to broaden the view of drug actions rooted in molecular interactions and advance the process of drug discovery. The aim of this work is to
Zheng Wu et al.
Clinical and experimental pharmacology & physiology, 41(7), 502-508 (2014-04-18)
The aim of the present study was to investigate the effects of salvianolic acid B on lipopolysaccharide (LPS)-induced disseminated intravascular coagulation (DIC) in rabbits. Continuous infusion of LPS was used to induce a DIC model in rabbits. Treatment with salvianolic
Mao-Liang Chen et al.
Immunopharmacology and immunotoxicology, 36(6), 390-396 (2014-10-15)
Tournefortia sarmentosa, a Chinese herbal medicine, is considered an antioxidant or a detoxicating agent. Recently T. sarmentosa has received attention for its effects on the immune response. Here we provide evidence that aqueous extract of T. sarmentosa can induce increased
Yong Fan et al.
Journal of pharmaceutical and biomedical analysis, 95, 213-219 (2014-04-02)
Sample extraction is the first challenge in analysis of herbal medicines (HMs). Numerous methods have been developed to improve extraction efficiency, use less solvent and short time. In this work, a tissue-smashing based ultra-rapid extraction (TSURE) method has been proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service