Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001550

Poly(vinyl alcohol)

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

polyvinyl alcohol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Poly(vinyl alcohol) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaobing Shen et al.
Journal of nanoscience and nanotechnology, 14(7), 5258-5265 (2014-04-25)
This study reports on the use of electrospun polyvinyl alcohol (PVA) nanofiber mats loaded with prazosin hydrochloride (PRH) as a transdermal drug delivery system, investigating the morphology of electrospun PVA nanofibers, the in vitro release characteristics of the drug from
Pedro Paulo Martins de Oliveira et al.
The Journal of thoracic and cardiovascular surgery, 147(4), 1405-1410 (2013-09-03)
Adhesions may increase the incidence of lethal complications of cardiac reoperations, which account for up to 20% of all open-heart surgeries. Herein, we describe the use of a polyvinyl alcohol membrane (PVAM) as a pericardial alternative and describe its performance
Yuichi Nogi et al.
International journal of systematic and evolutionary microbiology, 64(Pt 4), 1149-1153 (2014-01-11)
A polyvinyl-alcohol-degrading marine bacterium was isolated from plastic rope litter found in Tokyo Bay, Japan. The isolated strain, Zumi 95(T), was a Gram-reaction-negative, non-spore-forming and facultatively anaerobic chemo-organotroph. The major respiratory quinone was Q-10. The predominant fatty acids were C18 : 1ω7c
Lixin Su et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 73(1), 92-98 (2014-12-17)
To summarize the characteristics of Kasabach-Merritt syndrome (KMS) and to evaluate the therapeutic effect of drug therapy combined with transarterial embolization. From 2007 to 2011, 6 cases of KMS that underwent drug therapy and transarterial embolization were retrospectively analyzed, comprising
Yuming Dong et al.
Pharmaceutical development and technology, 19(8), 960-967 (2013-12-11)
High concentration of 5-amino salicylic acid (5-ASA) in the distal ileum and colon is necessary for the treatment of inflammatory bowel disease (IBD). The control of small molecules, drugs, released from a polymeric matrix remains a great challenge. To study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service