Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000677

Bromocriptine mesilate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Bromo-α-ergocryptine methanesulfonate salt, (+)-2-Bromo-12′-hydroxy-2′-(1-methylethyl)-5′-(2-methylpropyl)ergotaman-3′,6′-18-trione methanesulfonate salt, (+)-Bromocriptine methanesulfonate salt, Bromocriptine mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40BrN5O5 · CH4SO3
CAS Number:
Molecular Weight:
750.70
Beilstein:
4115238
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bromocriptine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CS(O)(=O)=O.[H][C@@]12Cc3c(Br)[nH]c4cccc(C1=CC(CN2C)C(=O)N[C@@]5(O[C@]6(O)N([C@@H](CC(C)C)C(=O)N7CCC[C@@]67[H])C5=O)C(C)C)c34

InChI

1S/C32H40BrN5O5.CH4O3S/c1-16(2)12-24-29(40)37-11-7-10-25(37)32(42)38(24)30(41)31(43-32,17(3)4)35-28(39)18-13-20-19-8-6-9-22-26(19)21(27(33)34-22)14-23(20)36(5)15-18;1-5(2,3)4/h6,8-9,13,16-18,23-25,34,42H,7,10-12,14-15H2,1-5H3,(H,35,39);1H3,(H,2,3,4)/t18?,23-,24+,25+,31-,32+;/m1./s1

InChI key

NOJMTMIRQRDZMT-NEKRQHSLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bromocriptine mesilate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V G Lyzogub et al.
Likars'ka sprava, (1-2)(1-2), 63-68 (2012-10-06)
In hypertensive patients with concomitant obesity observed decrease in activity of dopaminergic system in conjunction with increased activity of the renin-angiotensin-aldosterone system. Identified changes point to the advisability of appointing this group of patients the dopamine receptor agonist - bromocriptine-KV
Cory Sykora et al.
Respiratory physiology & neurobiology, 185(2), 257-264 (2012-10-17)
During and following hypoxic exposure young male hypothyroid hamsters treated with the dopamine D(2) receptor agonist bromocriptine increased breathing, while ventilation was depressed in bromocriptine-treated euthyroid hamsters. Moreover, D(2) receptor expression was increased in carotid bodies and striatum, but not
Yelena V Grinkova et al.
Biochemical and biophysical research communications, 430(4), 1223-1227 (2012-12-26)
The normal reaction mechanism of cytochrome P450 operates by utilizing two reducing equivalents to reduce atmospheric dioxygen, producing one molecule of water and an oxygenated product in an overall stoichiometry of 2 electrons:1 dioxygen:1 product. However, three alternate unproductive pathways
Bromocriptine-induced brainstem angulation in a patient with invasive prolactinoma.
Xiao Hui Lou et al.
The Journal of clinical endocrinology and metabolism, 98(3), 867-868 (2013-02-01)
J Michael Gaziano et al.
Journal of the American Heart Association, 1(5), e002279-e002279 (2013-01-15)
Bromocriptine-QR (a quick-release formulation of bromocriptine mesylate), a dopamine D2 receptor agonist, is a US Food and Drug Administrration-approved treatment for type 2 diabetes mellitus (T2DM). A 3070-subject randomized trial demonstrated a significant, 40% reduction in relative risk among bromocriptine-QR-treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service