Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000237

Perindopril impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Octahydroindole-2-carboxylic acid, (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO2
CAS Number:
Molecular Weight:
169.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

perindopril

manufacturer/tradename

EDQM

mp

259-260 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2N1

InChI

1S/C9H15NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h6-8,10H,1-5H2,(H,11,12)/t6-,7-,8-/m0/s1

InChI key

CQYBNXGHMBNGCG-FXQIFTODSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Perindopril impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liwei Liu et al.
ACS chemical biology, 10(3), 725-733 (2014-11-25)
Nodularia spumigena is a filamentous cyanobacterium that forms toxic blooms in brackish waters around the world through the production of the pentapeptide toxin nodularin. This cyanobacterium also produces large amounts of protease inhibitors belonging to the aeruginosin and spumigin families.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service