Skip to Content
MilliporeSigma
All Photos(1)

Documents

W324721

Sigma-Aldrich

Undecylenic acid

≥96%, FG

Synonym(s):

10-Undecenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3247
Colour Index Number:
42650
Beilstein:
1762631
EC Number:
Council of Europe no.:
689
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.039
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥96%

refractive index

n20/D 1.449 (lit.)

bp

137 °C/2 mmHg (lit.)

mp

23-25 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; sweet

SMILES string

OC(=O)CCCCCCCCC=C

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)

InChI key

FRPZMMHWLSIFAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Undecylenic acid is also referred to as 10-undecenoic acid. It exhibits anti-bacterial activity against various pathogenic fungi in the pH range of 4.5 to 6.0. It has fungicidal properties.

Application


  • Emulsion Liquid Membranes Based on Os-NP/n-Decanol or n-Dodecanol Nanodispersions for p-Nitrophenol Reduction.: This study explores the use of emulsion liquid membranes incorporating undecylenic acid for the reduction of p-nitrophenol. The findings highlight the potential of these membranes in environmental applications, specifically in the removal of harmful organic compounds from water (Pîrțac et al., 2024).

  • Retention behavior of nucleosides and nucleobases on a 3 μm undecylenic acid-functionalized silica column in per aqueous liquid chromatography and hydrophilic interaction liquid chromatography separation modes.: This article details the development of a novel undecylenic acid-functionalized silica column for improved separation of nucleosides and nucleobases. The column shows enhanced retention and selectivity, offering significant advancements in chromatographic techniques for biochemical analysis (Cheng et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of pH on the antifungal activity of undecylenic acid and its calcium salt.
Prince HN.
Journal of Bacteriology, 78(6), 788-788 (1959)
Rosario Pignatello et al.
Materials (Basel, Switzerland), 12(6) (2019-03-17)
A biodegradable poly(3-R-hydroxyalkanoate) synthesized by Pseudomonas mediterranea was investigated as a biomaterial to obtain colloidal drug delivery systems. Using a nanoprecipitation method, nanoparticles with a mean size of 155 nm and a negative surface charge were formed. They can be
Yusuke Yamamoto et al.
The Journal of toxicological sciences, 44(9), 585-600 (2019-09-03)
Amino acid derivative reactivity assay (ADRA) has previously been developed as an alternative method to direct peptide reactivity assay (DPRA) to evaluate key event 1 in skin sensitization mechanisms. However, when using alternative methods for skin sensitization, integrated approaches to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service