Skip to Content
MilliporeSigma
All Photos(4)

Documents

T1807

Sigma-Aldrich

L-(+)-Tartaric acid

BioXtra

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

vapor density

5.18 (vs air)

Quality Level

product line

BioXtra

Assay

≥98%

form

powder

autoignition temp.

797 °F

impurities

≤0.002% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

mp

170-172 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.002%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tartaric acid is a dicarboxylic acid and occurs as two enantiomers and an achiral meso compound. The L-(+)-tartaric acid is extensively found in nature. It is an acid present in fruits such as grapes and bananas. It possesses an astringent and citrus flavor.

Application

L-(+)-Tartaric acid has been used:
  • in filter-sterilized synthetic grape juice for feast fermentation
  • in crystallization experiment
  • to test the influence of stereospecific interactions between stereoisomers of tartaric acid and proteins during crystallogenesis

Biochem/physiol Actions

L-(+)-Tartaric acid serves as a donor ligand for biological processes. It is used as a food additive in candies and soft drinks to impart a sour taste.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4.16-human-environment interactions?Taste
Izawa K, et al.
Comprehensive Natural Products II, 631-671 (2010)
Acetogenin (Polypriopionate) Derived Auxiliaries: Tartaric Acid
Comprehensive Chirality (2012)
Higher Denticity Ligands
Comprehensive Coordination Chemistry II (2003)
Effects of protein purity and precipitant stereochemistry on the crystallization of thaumatin
Asherie N, et al.
Crystal Growth & Design, 8(12), 4200-4207 (2008)
Tartrate chirality determines thaumatin crystal habit
Asherie N, et al.
Crystal Growth & Design, 9(9), 4189-4198 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service