Skip to Content
MilliporeSigma
All Photos(1)

Documents

STS0022

Sigma-Aldrich

TERGITOL TMN-3

Non-ionic surfactant

Synonym(s):

alpha-[3,5-Dimethyl-1-(2-methylpropyl)hexyl]-omega-hydroxypolyloxy-1,2-ethanediyl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.28

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

viscosity

19 cP(25 °C)

transition temp

pour point -49 °C
cloud point insoluble

solubility

chlorinated solvents: soluble (and most polar and non-polar solvents and oils)
water: soluble (partially soluble)

density

0.928 g/mL at 20 °C

HLB

8.1

InChI

1S/C14H30O2/c1-11(2)8-13(5)10-14(9-12(3)4)16-7-6-15/h11-15H,6-10H2,1-5H3

InChI key

VKKFWRYCMZBXIG-UHFFFAOYSA-N

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service