Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01349

Sigma-Aldrich

Glutamyltyrosine

≥95% (HPLC)

Synonym(s):

N-alpha-L-Glutamyl-L-tyrosine, N-alpha-Glutamyltyrosine, L-Glu-L-tyr, L-Glutamyl-L-tyrosine, L-alpha-Glutamyl-L-tyrosine, Alpha-L-Glu-L-tyr, Alpha-Glu-tyr, Alpha-Glutamyltyrosine, a-L-Glu-L-tyr, a-Glu-tyr, a-Glutamyltyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O6
CAS Number:
Molecular Weight:
310.30
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.24

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

mp

186 °C

application(s)

(Metabolomics research)

storage temp.

2-8°C

SMILES string

N(C(Cc1ccc(cc1)O)C(=O)O)C(=O)C(N)CCC(=O)O

InChI

1S/C14H18N2O6/c15-10(5-6-12(18)19)13(20)16-11(14(21)22)7-8-1-3-9(17)4-2-8/h1-4,10-11,17H,5-7,15H2,(H,16,20)(H,18,19)(H,21,22)

InChI key

YSWHPLCDIMUKFE-UHFFFAOYSA-N

General description

Glutamyltyrosine is an incomplete breakdown product of protein digestion or protein catabolism. Some dipeptides are known to have physiological or cell-signaling effects although most are simply short-lived intermediates on their way to specific amino acid degradation pathways following further proteolysis.

Application

Glutamyltyrosine can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service