Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

S2054

Sigma-Aldrich

Sodium gluconate

meets USP testing specifications

Synonym(s):

D-Gluconic acid sodium salt, 2,3,4,5,6-Pentahydroxycaproic acid sodium salt, D-Gluconate sodium salt, Sodium D-gluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO7
CAS Number:
Molecular Weight:
218.14
Beilstein:
3919651
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

microbial

Quality Level

Agency

USP/NF
meets USP testing specifications

form

solid

color

beige

useful pH range

6.5 - 7.5

mp

170-175  °C

solubility

590 g/L at 25 °C

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

UPMFZISCCZSDND-JJKGCWMISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium gluconate has been used as a component of recording buffer used in two-electrode voltage-clamp (TEVC) recording in Xenopus laevis oocytes. It has also been used as a control for sodium.

Biochem/physiol Actions

Ingestion of sodium gluconate is known to stimulate the production of intestinal butyrate. It is widely used in food, pharmaceutical paper and textile industry. It acts as a chelating agent. Sodium gluconate serves as a detergent in bottle washing formulation.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Two-electrode voltage-clamp recordings in Xenopus laevis oocytes: reconstitution of abscisic acid activation of SLAC1 anion channel via PYL9 ABA receptor
Wang C, et al.
Bio-protocol, 7(2) (2017)
Dietary sodium gluconate protects rats from large bowel cancer by stimulating butyrate production
Kameue C, et al.
The Journal of Nutrition, 134(4), 940-944 (2004)
Influence of nitrate and nitrite on thyroid hormone responsive and stress-associated gene expression in cultured Rana catesbeiana tadpole tail fin tissue
Hinther A, et al.
Frontiers in Genetics, 3, 51-51 (2012)
Gluconic acid: Properties, applications and microbial production.
Ramachandran S, et al.
Food Technology and Biotechnology, 44(2) (2006)
Elena von Molitor et al.
Cell calcium, 87, 102164-102164 (2020-02-06)
Bridging the gap between two-dimensional cell cultures and complex in vivo tissues, three-dimensional cell culture models are of increasing interest in the fields of cell biology and pharmacology. However, present challenges hamper live cell imaging of three-dimensional cell cultures. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service