Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR3738

Supelco

Propionic acid

certified reference material, pharmaceutical secondary standard

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
Beilstein:
506071
MDL number:
UNSPSC Code:
12164502

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1572208

vapor density

2.55 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

autoignition temp.

955 °F

expl. lim.

12.1 %

packaging

pkg of 500MG

refractive index

n20/D 1.386 (lit.)

bp

141 °C (lit.)

mp

−24-−23 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service