Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR3118

Supelco

Propofol Related Compound C

new

pharmaceutical secondary standard, certified reference material

Synonym(s):

2,6-Diisopropylphenyl isopropyl ether, 2-(1-Methylethoxy)-1,3-bis(1-methylethyl)benzene, Propofol isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
MDL number:

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1572558

API family

propofol

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

application(s)

pharmaceutical

InChI

1S/C15H24O/c1-10(2)13-8-7-9-14(11(3)4)15(13)16-12(5)6/h7-12H,1-6H3

InChI key

APXUVWNNDMEOEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service