Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0950000

Phenoxyethanol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Phenoxyethanol, Ethylene glycol monophenyl ether, Phenylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1364011
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenoxyethanol

manufacturer/tradename

EDQM

bp

244-246 °C

mp

11-13 °C

density

1.107 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OCCOc1ccccc1

InChI

1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

QCDWFXQBSFUVSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenoxyethanol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Scognamiglio et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S244-S255 (2011-11-01)
A toxicologic and dermatologic review of 2-phenoxyethanol when used as a fragrance ingredient is presented. 2-Phenoxyethanol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a primary alcohol. The AAAs are a structurally diverse class of
Angela E Aakhus et al.
Dermatitis : contact, atopic, occupational, drug, 22(3), 127-140 (2011-05-17)
Methyldibromoglutaronitrile/phenoxyethanol (Euxyl K 400) is a preservative found in both personal care products and industrial sources. Although Euxyl K 400 initially appeared to have low sensitizing potential, increased prevalence of contact allergy to Euxyl K 400 led to regulatory intervention.
T W Chew et al.
Oncogene, 33(21), 2717-2727 (2013-07-03)
RAS and Rho small GTPases are key molecular switches that control cell dynamics, cell growth and tissue development through their distinct signaling pathways. Although much has been learnt about their individual functions in both cell and animal models, the physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service