Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2831

Vickers hardness of ceramics and hardmetals

NIST® SRM® 2831

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

disc

packaging

pkg of each

manufacturer/tradename

NIST®

General description

Each unit of SRM 2831 contains 25 mm D × 9.5 mm disk with a nominal hardness of ~15.0 GPa (1530kgf/mm2), packaged in a wooden box. Every unit is certified individually and features a unique serial number, which is inscribed on the opposite (bottom)face of the disk in the format of the letter W followed by a hyphen and the disk number.

SRM 2831_cert

SRM 2831_SDS

Application

Vickers hardness of ceramicsand hard metals standard reference material (SRM) is intended for calibrating all hardness and microhardness testing machines. It involves creating a Vickers indentation and subsequently measuring it using a microscope.

Features and Benefits

  • Accompanied with certified values for individual diagonal lengths, average diagonal length, and average Vickers hardness are provided.
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service