Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1980

Positive electrophoretic mobility standard

NIST® SRM® 1980

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

suspension

packaging

pkg of 500_mg/L

manufacturer/tradename

NIST®

application(s)

general analytical

General description

Each unit of SRM 1980 contains a single 60 cm³ polyethylene bottle filled with 40 cm³ of suspension, which should be diluted before analysis. The suspension contains a 500 mg/L goethite (α-FeOOH) saturated with 100 μmol/g phosphate in a 5 × 10⁻² mol/L sodium perchlorate electrolyte solution, with a pH of 2.5. For more information, please refer to the COA and SDS.

SRM 1980_cert

SRM 1980_SDS

Application

The Standard Reference Material (SRM) is primarily intended for use in evaluating equipment and validating methodologies for measuring electrophoretic mobility.

Features and Benefits

  • NIST certified value are metrologically traceable to the SI unit for meter, volt, and time and expressed as µm·cm/V·s).
  • Expiration details, instructions for storage and use are provided on the NIST certificate.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service