Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1951C

Lipids in frozen human serum

NIST® SRM® 1951c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

pkg of 4 vials (2 ea conc)

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

General description

A unit of SRM1951C consists of four vials of frozen human serum, two vials each at two different analyte concentration levels. Each bottle contains approximately 1 mL of serum.

NIST1951C_Cert

NIST1951C_SDS

Application

The lipids in frozen human serum are a standard reference material (SRM) that is used for evaluating the accuracy of clinical procedures for the determination of total cholesterol, high-density lipoprotein (HDL)-cholesterol, low-density lipoprotein (LDL)-cholesterol, and total glycerides in human serum. It is also used for validating working or secondary reference materials.

Other Notes

  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate.
  • Notes on biomaterials, disposal, and transport are documented in SDS.
Certified for the analytes listed below.
Total Cholesterol, Total Glycerides

Reference/Informational value is provided for the following analytes.
HDL-Cholesterol, LDL-Cholesterol, Total Cholesterol

See certificate for values and more details at nist.gov/SRM.

Preparation Note

  • The serum is shipped frozen on dry ice and after receiving it should be frozen until ready for use.
  • Before use, the bottles must be thawed to room temperature.
  • The certified concentrations apply only to serum thawed to room temperature, 20 °C to 25 °C.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The extended lipid panel assay: a clinicallydeployed high-throughput nuclear magnetic resonance method for the simultaneous measurement of lipids and Apolipoprotein B
Erwin G, et al.
Adv. in Lipids Research (2020)
Beatriz Jiménez et al.
Analytical chemistry, 90(20), 11962-11971 (2018-09-14)
We report an extensive 600 MHz NMR trial of quantitative lipoprotein and small-molecule measurements in human blood serum and plasma. Five centers with eleven 600 MHz NMR spectrometers were used to analyze 98 samples including 20 quality controls (QCs), 37

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service