Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM633

Supelco

VOCs - Loamy Sand 4

pkg of 30 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 30 g

manufacturer/tradename

RTC CRM633-030

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

shipped in

wet ice

storage temp.

-10 to -25°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Acetone
    Benzene
    Bromobenzene
    Bromodichloromethane
    2-Butanone
    Carbon tetrachloride
    Chlorobenzene
    Chloroform
    Chloromethane
    Dibromochloromethane
    Dibromomethane
    1,2-Dibromo-3-chloropropane
    1,2-Dibromoethane
    1,2-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    Ethylbenzene
    Bromomethane
    Methyl tert-butyl ether solution
    Styrene
    1,1,2,2-Tetrachloroethane
    Toluene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    1,2,3-Trichloropropane
    1,2,4-Trimethylbenzene
    Mesitylene
    o-Xylene
    See All (30)

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service