Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM608

Supelco

VOCs - Sandy Loam 1

pkg of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 25 g

manufacturer/tradename

RTC CRM608-025

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The following elements are not certified and are included for information only:
  • Acetonitrile
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Acetone
    Acetonitrile
    Benzene
    Bromobenzene
    Bromodichloromethane
    Bromoform
    2-Butanone
    Carbon tetrachloride
    Chlorobenzene
    Chloroethane
    Chloroform
    1,2-Dibromo-3-chloropropane
    Dibromochloromethane
    1,2-Dibromoethane
    Dibromomethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    trans-1,2-Dichloroethylene
    Ethylbenzene
    2-Hexanone
    Cumene
    Bromomethane
    Chloromethane
    4-Methyl-2-pentanone
    tert-Butyl methyl ether
    Styrene
    1,1,1,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichlorofluoromethane
    1,2,3-Trichloropropane
    Mesitylene
    Vinyl acetate
    Vinyl chloride
    o-Xylene
    See All (46)

Hazard Statements

Precautionary Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service