Skip to Content
MilliporeSigma
All Photos(1)

Documents

COND2060

Supelco

Conductivity Standard - 2060 μmhos/cm

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-2060-0.500

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This ready-to-use certified reference material (CRM) is manufactured and certified according to ISO 17034 and ISO/IEC 17025. The specific conductance value of 2060 ± 75 µmhos/ cm is traceable to SI and higher order standards from National Institute of Standards and Technology (NIST). The certified value is based on purity determination by mass balance and gravimetrically prepared value.

All information regarding packaging, uncertainty, homogeneity and stability is documented in the certificate of analysis (CoA).

Application

Conductivity calibration standard solution for monitoring and checking of conductivity meters.

Used as a standard in the construction of electrical conductivity sensors for detection and chemical mapping of ammonia release.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Developing a network of inexpensive electrical conductivity sensors to map ammonia release
Collins MD, et al.
Chemical, Biological, Radiological, Nuclear, and Explosives (CBRNE) Sensing XIII, 12116, 133-139 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service