Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR614

Polychlorodibenzo-p-dioxins (PCDDs) and polychlorodibenzofurans (PCDFs) solution

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Components

The nine solutions of natural and labeled PCDD and PCDF congeners in n-nonane are presented in brown glass ampoules sealed under helium gas. Ampoules S1 to S7 contain about 1 mL solution and ampoule S8 about 0.5 mL. Solutions S1-S8 are supplied in one set together with a non-certified solution S9 intended to verify performance of the chromatographic separation system. The set consists of 11 ampoules, one of S2, S4-S9 and two of S1 and S3.

Analysis Note

For more information please see:
BCR614

Legal Information

BCR is a registered trademark of European Commission

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service