Skip to Content
MilliporeSigma
All Photos(3)

Documents

B0426

Sigma-Aldrich

Bismuth subnitrate

meets USP testing specifications

Synonym(s):

Bismuth(III) subnitrate, Bismuth(III) nitrate basic, Bismuth(III) oxynitrate, Bismuthyl nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi5O(OH)9(NO3)4
CAS Number:
Molecular Weight:
1461.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

powder

application(s)

pharmaceutical (small molecule)

SMILES string

O[Bi]=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O.O[Bi](O)O[N+]([O-])=O

InChI

1S/5Bi.4NO3.9H2O.O/c;;;;;4*2-1(3)4;;;;;;;;;;/h;;;;;;;;;9*1H2;/q+1;4*+3;4*-1;;;;;;;;;;/p-9

InChI key

OAVDTZZNYFIIGB-UHFFFAOYSA-E

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z M Orziev et al.
Klinicheskaia meditsina, 81(6), 48-49 (2003-07-24)
46 patients having exacerbation of duodenal ulcer in the presence of Helicobacter pylori (HP) were studied for efficacy of HP eradication treatment which comprised omeprazole (40 mg/day), colloid bismuth subcitrate (480 mg/day) and metronidazole (1000 mg/day) for 2 weeks. According
Meredin Stoltenberg et al.
Journal of applied toxicology : JAT, 22(2), 111-115 (2002-03-29)
The consumption of bismuth is increasing and knowledge of the potential teratogenic and reproductive damage of bismuth exposure is fragmentary. In the present study we used autometallography (AMG)--the histochemical silver amplification technique--to trace bismuth in the testis and pituitary glands
K I Parker et al.
Journal of dairy science, 90(1), 207-218 (2006-12-22)
This study investigated the effect in heifers of infusion of a bismuth subnitrate teat-canal sealant and bacterial intramammary infection (IMI) precalving on prevalence of postcalving IMI and incidence of clinical mastitis in the first 2 wk postcalving. Glands (n =
Michael D Levitt et al.
Journal of applied physiology (Bethesda, Md. : 1985), 92(4), 1655-1660 (2002-03-16)
Colonic bacteria produce hydrogen sulfide, a toxic compound postulated to play a pathogenetic role in ulcerative colitis. Colonic sulfide exposure has previously been assessed via measurements of fecal sulfide concentration. However, we found that <1% of fecal sulfide of rats
M Stoltenberg et al.
European journal of histochemistry : EJH, 51(1), 53-57 (2007-06-06)
Bismuth - sulphur quantum dots can be silver enhanced by autometallography (AMG). In the present study, autometallographic silver enhanced bismuth-sulphur nanocrystals were isolated from unfixed cryo-sections of kidneys and livers of rats exposed to bismuth (Bi207) subnitrate. After being subjected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service