Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96373

Supelco

Glyceryl trilinoleate

analytical standard

Synonym(s):

1,2,3-Tri-(cis,cis-9,12-octadecadienoyl)glycerol, 1,2,3-Trilinoleoylglycerol, Glycerol trilinoleate, Trilinolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H98O6
CAS Number:
Molecular Weight:
879.38
Beilstein:
1718698
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)
≥97.5% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.478-1.480

density

0.925 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

functional group

ester

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C57H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16-21,25-30,54H,4-15,22-24,31-53H2,1-3H3/b19-16-,20-17-,21-18-,28-25-,29-26-,30-27-

InChI key

HBOQXIRUPVQLKX-BBWANDEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glyceryl trilinoleate may be used as one of the standard for evaluating the kinetics of blackcurrant seed oil lipolysis in supercritical carbon dioxide by reversed phase high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC in reversed phase mode: Tool for investigation of kinetics of blackcurrant seed oil lipolysis in supercritical carbon dioxide.
Bartlova M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 839(1-2), 80-84 (2006)
Chandra Durgampudi et al.
The American journal of pathology, 184(6), 1773-1784 (2014-05-24)
Obese patients have worse outcomes during acute pancreatitis (AP). Previous animal models of AP have found worse outcomes in obese rodents who may have a baseline proinflammatory state. Our aim was to study the role of acute lipolytic generation of
Alexandra Paul et al.
Biophysical journal, 116(12), 2346-2355 (2019-06-04)
Lipid droplets (LDs), present in many cell types, are highly dynamic organelles that store neutral lipids, primarily triacylglycerols (TAGs). With the discovery of new LD functions (e.g., in immune response, protein clearage, and occurrence with disease), new methods to study
Veerle W Daniëls et al.
PloS one, 9(9), e106913-e106913 (2014-09-13)
Increased lipogenesis is a hallmark of a wide variety of cancers and is under intense investigation as potential antineoplastic target. Although brisk lipogenesis is observed in the presence of exogenous lipids, evidence is mounting that these lipids may adversely affect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service