Skip to Content
MilliporeSigma
All Photos(1)

Documents

96328

Sigma-Aldrich

tert-Butylamine hydrochloride

≥98.0% (AT)

Synonym(s):

2-Amino-2-methylpropane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNH2 · HCl
CAS Number:
Molecular Weight:
109.60
Beilstein:
3905687
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥98.0% (AT)

form

powder

impurities

<0.5% water

SMILES string

Cl.CC(C)(C)N

InChI

1S/C4H11N.ClH/c1-4(2,3)5;/h5H2,1-3H3;1H

InChI key

DLDIDQIZPBIVNQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Ekkati et al.
Amino acids, 38(3), 747-751 (2009-04-01)
N-Acetyl-AA(amino acid)-NHtBu derivatives of all 20 naturally occurring amino acids have been synthesized. Syntheses were performed via solution-phase methodology with yields that allow for access to gram quantities of substrates, in most cases. Syntheses include the coupling of a hindered
Marina Bicchieri et al.
Analytical and bioanalytical chemistry, 402(4), 1517-1528 (2011-07-14)
The Italian dialect poet Cesare Pascarella travelled all around the world, noting down in notebooks his keen and caustic observations, and drawing sketches that are a visual reportage of his journeys. The sketches were mounted as a random collage over
Kana Sadaoka et al.
Bioorganic & medicinal chemistry, 19(13), 3901-3905 (2011-06-15)
Reduction of the 7-formyl groups in chlorophyll (Chl) b and its demetalated compound pheophytin (Phe) b was kinetically analyzed by using tert-butylamine-borane complex (t-BuNH(2)·BH(3)), and was compared with that of the 3-formyl groups in Chl d and Phe d. Reduction
Z L Bandi
Clinical chemistry, 27(10), 1676-1681 (1981-10-01)
We find that 2 to 6 mmol of carbon dioxide per liter (mean: 4.1 mmol/L) is lost during routine laboratory processing of patients' serum samples after centrifugation. Additional CO2 may be lost if evacuated blood-collection tubes are not filled completely
M S Suleiman et al.
Journal of pharmaceutical and biomedical analysis, 8(4), 321-327 (1990-01-01)
A new salt of ibuprofen was prepared by reaction with t-butylamine; its formation was confirmed by IR and 1H-NMR spectroscopy. The salt was characterized by thermoanalytical, X-ray powder diffraction and solubility studies. The salt was found to be 1.5 times

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service