Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

92648

Supelco

Betulin

analytical standard

Synonym(s):

Betulinic alcohol, Betulinol, Betulol, Lup-20(29)-ene-3β,28-diol, Trochol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O2
CAS Number:
Molecular Weight:
442.72
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

256-257 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@]12CC[C@]3([H])[C@@]4(C)CC[C@H](O)C(C)(C)[C@]4([H])CC[C@@]3(C)[C@]1(C)CC[C@@]5(CO)CC[C@@H](C(C)=C)[C@]25[H]

InChI

1S/C30H50O2/c1-19(2)20-10-15-30(18-31)17-16-28(6)21(25(20)30)8-9-23-27(5)13-12-24(32)26(3,4)22(27)11-14-29(23,28)7/h20-25,31-32H,1,8-18H2,2-7H3/t20-,21+,22-,23+,24-,25+,27-,28+,29+,30+/m0/s1

InChI key

FVWJYYTZTCVBKE-ROUWMTJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betulin is a naturally occurring pentacyclic triterpene alcohol with a lupane skeleton, found in large quantities in the bark of white birch. It may possess anticancer and antiviral properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: salvia sambucus

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycopolymers bearing galactose and betulin: Synthesis, encapsulation, and lectin recognition
Ma Z, et al.
Biomacromolecules, 18, 3812-3818 (2017)
Miroslav Pospíšil et al.
Journal of molecular modeling, 18(1), 367-376 (2011-05-04)
Ab initio and molecular simulation methods were used in calculations of the neutral individual betulin molecule, and molecular simulations were used to optimize the betulin molecule immersed in various amounts of water. Individual betulin was optimized in different force fields
Cristina A Dehelean et al.
Journal of biomedical nanotechnology, 9(4), 577-589 (2013-04-30)
Betulin (Bet), the main component of birch tree bark, has been recently reported to exert anticancer activity in several cell lines; however the underlying mechanisms are only partially elucidated. The aims of the present work were to assess the in
Codruta M Soica et al.
Natural product research, 26(10), 968-974 (2011-05-21)
Betulin and betulinic acid are pentacyclic triterpenes present in the bark of the birch tree and other vegetal sources. Quantitatively, in birch bark betulin is more significant than betulinic acid; therefore, birch can be a large and feasible source of
Sayan Chowdhury et al.
Molecular pharmacology, 80(4), 694-703 (2011-07-14)
Toward developing antileishmanial agents with mode of action targeted to DNA topoisomerases of Leishmania donovani, we have synthesized a large number of derivatives of betulin. The compound, a natural triterpene isolated from the cork layer of Betula spp. plants exhibits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service