Skip to Content
MilliporeSigma
All Photos(1)

Documents

92615

Supelco

Tin Standard for ICP

TraceCERT®, 1 g/L Sn in hydrochloric acid (nominal concentration)

Synonym(s):

Tin Calibration Solution for ICP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Sn in hydrochloric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Tin ICP standard is a suitable calibration standard for elemental analysis by InductivelyCoupled Plasma (ICP) technique.

Analysis Note

1 g/L Sn in 10% hydrochloric acid, prepared with high purity tin metal, HCl and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service