Skip to Content
MilliporeSigma
All Photos(1)

Documents

90548

Supelco

Tetrathiafulvalene 7,7,8,8-tetracyanoquinodimethane salt

≥97.0% (CHNS)

Synonym(s):

7,7,8,8-Tetracyanoquinodimethane Tetrathiafulvalene salt, TCNQ-TTF, TTF-TCNQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4S4 · C12H4N4
CAS Number:
Molecular Weight:
408.54
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

Assay

≥97.0% (CHNS)

form

solid

mp

~230 °C (dec.)

SMILES string

S1C=CS\C1=C2/SC=CS2.N#C\C(C#N)=C3\C=CC(\C=C3)=C(\C#N)C#N

InChI

1S/C12H4N4.C6H4S4/c13-5-11(6-14)9-1-2-10(4-3-9)12(7-15)8-16;1-2-8-5(7-1)6-9-3-4-10-6/h1-4H;1-4H

InChI key

OIXMVDHMELKBDX-UHFFFAOYSA-N

General description

TTF-TCNQ is an organic metal-like charge transfer complex, which is mostly used as an electrode.
Visit our Sensor Applications portal to learn more.

Application

TTF-TCNQ has been used in sensor fabrication during amperometric lactate biosensor development for qualitative analysis of lactate accumulation in ischemic myocardium patients.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Kataky et al.
Talanta, 56(3), 451-458 (2008-10-31)
A format for a disposable screen-printed biosensor was investigated for monitoring formaldehyde. The screen-printed sensor format comprised a working electrode (WE) modified with platinised carbon, a new lipophilic tetrathiafulvalene-tetracyanoquinodimethane (TTF-TCNQ) salt as mediator, a plasticised polyurethane membrane (TECOFLEX SG80, PU)
Seokmin Jeon et al.
ACS nano, 10(8), 7821-7829 (2016-07-28)
Bulk molecular ionic solids exhibit fascinating electronic properties, including electron correlations, phase transitions, and superconducting ground states. In contrast, few of these phenomena have been observed in low-dimensional molecular structures, including thin films, nanoparticles, and molecular blends, not in the
Maxime Leroux et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(22), 10691-10697 (2019-05-16)
With the discovery of charge-density waves (CDWs) in most members of the cuprate high-temperature superconductors, the interplay between superconductivity and CDWs has become a key point in the debate on the origin of high-temperature superconductivity. Some experiments in cuprates point
Fariba Mashayekhi Mazar et al.
Advanced materials (Deerfield Beach, Fla.), 31(32), e1901677-e1901677 (2019-06-20)
Untethered actuation is important for robotic devices to achieve autonomous motion, which is typically enabled by using batteries. Using enzymes to provide the required electrical charge is particularly interesting as it will enable direct harvesting of fuel components from a
A conducting salt-based amperometric biosensor for measurement of extracellular lactate accumulation in ischemic myocardium.
Marzouk SA
Analytical Chemistry, 69(14), 2646-2652 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service