Skip to Content
MilliporeSigma
All Photos(1)

Documents

86874

Sigma-Aldrich

Tetrabutylammonium hexafluorophosphate

purum, ≥98.0% (CHN)

Synonym(s):

TBAPF6, Tetra-n-butylammonium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(PF6)
CAS Number:
Molecular Weight:
387.43
Beilstein:
4056584
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (CHN)

form

powder

impurities

≤0.3% halogenides (as Br)
≤1% water

mp

244-246 °C (lit.)

SMILES string

F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.F6P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-7(2,3,4,5)6/h5-16H2,1-4H3;/q+1;-1

InChI key

BKBKEFQIOUYLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium hexafluorophosphate (TBAPF6) or TBAHP can be used:
  • As the supporting electrolyte in cyclic voltammetric studies.
  • As a reactant in the synthesis of fluorescent nanofibrous sensing films for detecting nitro-explosive vapors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Signal-Amplifying Fluorescent Nanofibers for Naked-Eye-Based Ultrasensitive Detection of Buried Explosives and Explosive Vapors
Wang Y, et al.
Advances in Functional Materials, 22(17), 3547-3555 (2012)
Switching on elusive organometallic mechanisms with photoredox catalysis
Terrett JA, et al.
Nature, 524(7565), 330-330 (2015)
Tetrabutylammonium hexafluorophosphate and 1-ethyl-3-methyl imidazolium hexafluorophosphate ionic liquids as supporting electrolytes for non-aqueous vanadium redox flow batteries
Zhang D, et al.
Journal of Power Sources, 203(7565), 201-205 (2012)
Sonia Bruña et al.
Dalton transactions (Cambridge, England : 2003), 43(35), 13187-13195 (2014-07-17)
Reaction of Fe3(CO)12 with 1,2-dithiolene HSC6H2Cl2SH affords a mixture of complexes [Fe2(CO)6(μ-SC6H2Cl2S)] 1, [Fe2(SC6H2Cl2S)4] 2 and [Fe3(CO)7(μ3-SC6H2Cl2S)2] 3. In the course of the reaction the trimetallic cluster 3 is first formed and then converted into the known dinuclear compound 1
Steven Hananouchi et al.
Dalton transactions (Cambridge, England : 2003), 43(48), 17991-18000 (2014-10-30)
Isostructural vanadium, niobium and tantalum complexes of bis(3,5-di-tert-butyl-2-phenol)amine ([ONO]H3), were prepared and characterized to evaluate the impact of the metal ion on redox-activity of the ligand platform. New vanadium and niobium complexes with the general formula, [ONO]MCl2L (M = V

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service