Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84605

Millipore

SD Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

composition

agar, 20 g/L
ammonium sulfate, 5 g/L
biotin, 0.02 mg/L
boric acid, 0.5 mg/L
calcium chloride, 0.1 g/L
calcium pantothenate, 2 mg/L
copper sulfate, 0.04 mg/L
dextrose, 20 g/L
dipotassium hydrogen phosphate, 0.15 g/L
ferric chloride, 0.2 mg/L
folic acid, 0.002 mg/L
inositol, 10 mg/L
magnesium sulfate, 0.5 g/L
manganese sulfate, 0.4 mg/L
niacin, 0.4 mg/L
PABA, 0.2 mg/L
potassium dihydrogen phosphate, 0.85 g/L
potassium iodide, 0.1 mg/L
pyridoxine HCl, 0.4 mg/L
riboflavin, 0.2 mg/L
sodium chloride, 0.1 g/L
sodium molybdate, 0.2 mg/L
thiamine HCl, 0.4 mg/L
zinc sulfate, 0.4 mg/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

5.4±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

nonselective for Candida spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.

Application

Synthetic medium for the growth of yeast for molecular biology purposes, Sherman (1991).

Preparation Note

Dissolve 46.7 g in 1 litre distilled water and sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Getting started with yeast.
F Sherman
Methods in enzymology, 194, 3-21 (1991-01-01)

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service