Skip to Content
MilliporeSigma

80038

Melamine-(triamine-15N3)

analytical standard

Synonym(s):

1,3,5-Triazine-2,4,6-triamine-15N3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H615N3N3
CAS Number:
Molecular Weight:
129.10
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+3

storage temp.

2-8°C

SMILES string

[15NH2]c1nc([15NH2])nc([15NH2])n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i4+1,5+1,6+1

InChI key

JDSHMPZPIAZGSV-VMGGCIAMSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

15N enrichment
minimum 80 atom% 15N (triamine)
minimum 20 atom% 15N (triazine)


Still not finding the right product?

Explore all of our products under Melamine-(triamine-15N3)


flash_point_c

300 °C - closed cup

pictograms

Health hazard

signalword

Warning

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

target_organs

Urinary tract

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

572.0 °F - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library