Skip to Content
MilliporeSigma

79997

Azodicarboxamide

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2CON=NCONH2
CAS Number:
Molecular Weight:
116.08
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
204-650-8
Beilstein/REAXYS Number:
1704003
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

96.0-104.0% (RT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

220-225 °C (dec.) (lit.)

application(s)

environmental

format

neat

SMILES string

NC(=O)\N=N\C(N)=O

InChI

1S/C2H4N4O2/c3-1(7)5-6-2(4)8/h(H2,3,7)(H2,4,8)/b6-5+

InChI key

XOZUGNYVDXMRKW-AATRIKPKSA-N

General description

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)


Still not finding the right product?

Explore all of our products under Azodicarboxamide


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

supp_hazards

Storage Class

4.1A - Other explosive hazardous materials

wgk

WGK 1

flash_point_f

>392.0 °F

flash_point_c

> 200 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library