Skip to Content
MilliporeSigma

77091

Palladium Standard for ICP

TraceCERT®, 1 g/L Pd in hydrochloric acid (nominal concentration)

Synonym(s):

ICP Palladium Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12141733
NACRES:
NA.24
eCl@ss:
38191001
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

concentration

1 g/L Pd in hydrochloric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Palladium standard for ICP can be used as internal standard in the quantitative analysis of various types of solids via laser ablation technique in combination with inductively coupled plasma-mass spectrometry (LA-ICPMS).

Analysis Note

1 g/L Pd in 5% hydrochloric acid (contains HNO3 traces), prepared with high purity Pd metal, HCl, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Palladium Standard for ICP


pictograms

CorrosionEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library