Skip to Content
MilliporeSigma
All Photos(1)

Documents

76731

Supelco

Density Standard 870 kg/m3

H&D Fitzgerald Ltd. Quality

Synonym(s):

Mineral oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

H&D Fitzgerald Ltd. Quality
analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

H&D Fitzgerald Ltd.

refractive index

n20/D 1.467 (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
pharmaceutical (small molecule)

format

mixture

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

Application

This density CRM is used for the calibration of all types of laboratory density meters in the test temperature range of 10 to 50 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.010 Kg/m3
  • Provided in a sealed glass ampoule of 10 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service