Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38788

Supelco

Copper(II) Ionophore V

Selectophore, function tested

Synonym(s):

2-(1,4,8,11-Tetrathiacyclotetradec-6-yloxy)hexanoic acid, 2-(3,6,10,13-Tetrathiacyclotetradec-1-oxy)hexanoic acid, TTCTOHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O3S4
CAS Number:
Molecular Weight:
398.67
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

quality

function tested

SMILES string

CCCCC(OC1CSCCSCCCSCCSC1)C(O)=O

InChI

1S/C16H30O3S4/c1-2-3-5-15(16(17)18)19-14-12-22-10-8-20-6-4-7-21-9-11-23-13-14/h14-15H,2-13H2,1H3,(H,17,18)

InChI key

ZIRYKADCKCUEGL-UHFFFAOYSA-N

General description

Copper(II) Ionophore V is a calboxylated thiacrown ether.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of thiacrown ether carboxylic acids and their characteristics as extractants for metal ions.
Saito, Keiitsu, et al.
Analytica Chimica Acta, 299, 137-144 (1994)
Copper(II)-selective electrodes based on a novel charged carrier and preliminary application of field-effect transistor type checker
Wakida, S., et al.
Sensors and Actuators B, Chemical, 130, 187-192 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service