Skip to Content
MilliporeSigma
All Photos(1)

Documents

38390

Millipore

Preston Selective Supplement

suitable for microbiology

Synonym(s):

Campylobacter Selective Supplement IV, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

Application

This supplement is recommended for selective isolation of Campylobacter species.

Components

Composition:
Per vial sufficient for 500 ml medium
Polymyxin B 2500 IU
Rifampicin 5 mg
Trimethoprim lactate 5 mg
Amphotericin B 5 mg

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service