Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37886

Supelco

Buprofezin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23N3OS
CAS Number:
Molecular Weight:
305.44
Beilstein:
8324923
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)N1C(=O)N(CS\C1=N\C(C)(C)C)c2ccccc2

InChI

1S/C16H23N3OS/c1-12(2)19-14(17-16(3,4)5)21-11-18(15(19)20)13-9-7-6-8-10-13/h6-10,12H,11H2,1-5H3/b17-14+

InChI key

PRLVTUNWOQKEAI-SAPNQHFASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Buprofezin is an insect growth regulator insecticide. Its mode of action involves inhibition of chitin biosynthesis and is also known to inhibit molting against coleopteran, hemipteran insects and acarina.

Application

Buprofezin may be used as a reference standard for the determination of buprofezin in rice and fruits by high-performance liquid chromatography followed by liquid chromatography-mass spectrometry confirmation.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugo R Monteiro et al.
Chemosphere, 221, 324-332 (2019-01-15)
Acute and chronic toxicity of the formamidine pesticide amitraz to the midge Chironomus riparius was assessed using conventional ecotoxicological tests and biochemical approaches (biomarkers). Amitraz is mainly used as an ectoparasiticide in veterinary medicine, but also in agriculture and apiculture.
Muhammad D Gogi et al.
Pest management science, 62(10), 982-990 (2006-07-25)
Field efficacies of two insect growth regulators (IGRs) at two recommended application rates, buprofezin at 370 and 555 g AI ha(-1) and lufenuron at 37 and 49 g AI ha(-1), were determined against the sweet potato whitefly, Bemisia tabaci (Gennadius)
E E Grafton-Cardwell et al.
Journal of economic entomology, 99(3), 733-744 (2006-07-04)
Portions of two commercial citrus orchards were treated for two consecutive years with buprofezin or three consecutive years with pyriproxyfen in a replicated plot design to determine the long-term impact of these insect growth regulators (IGRs) on the San Joaquin
Chao Li et al.
Journal of agricultural and food chemistry, 60(10), 2531-2537 (2012-02-18)
A buprofezin-degrading bacterium, YL-1, was isolated from rice field soil. YL-1 was identified as Rhodococcus sp. on the basis of the comparative analysis of 16S rDNA sequences. The strain could use buprofezin as the sole source of carbon and nitrogen
Yanhua Wang et al.
Pest management science, 64(10), 1050-1056 (2008-05-29)
Buprofezin has been used for many years to control Nilaparvata lugens (Stål). Assessment of susceptibility change in the insect is essential for maintaining control efficiency and resistance management. Eleven-year surveys showed that most field populations were susceptible before 2004. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service