Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36575

Supelco

Gibberellic acid

PESTANAL®, analytical standard

Synonym(s):

GA3, Gibberellin A3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O6
CAS Number:
Molecular Weight:
346.37
Beilstein:
54346
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

90-100% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@]12[C@@H](O)C=C[C@@]3(OC1=O)[C@@H]4CC[C@]5(O)C[C@]4(CC5=C)[C@H]([C@H]23)C(O)=O

InChI

1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16-,17+,18+,19-/m1/s1

InChI key

IXORZMNAPKEEDV-OBDJNFEBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gibberellic acid is an endogenous plant growth regulator
Gibberellic acid is a plant growth regulator, which can find applications in agriculture, to improve germination, overall development of the plant, increase the productivity and also the quality of food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Phytohormone that controls important aspects of plant growth: germination, elongation, and flowering. The effects are mediated by inducing degradation of the DELLA repressor protein, and probably other minor signalling pathways.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?-Polyglutamic acid/chitosan nanoparticles for the plant growth regulator gibberellic acid: Characterization and evaluation of biological activity
Pereira S.E.A, et al.
Carbohydrate Polymers, 157, 1862-1873 (2017)
Chitosan nanoparticles as carrier systems for the plant growth hormone gibberellic acid
Pereira S.E.A, et al.
Colloids and Surfaces. B, Biointerfaces, 150, 141-152 (2017)
Shulan Sun et al.
Journal of experimental botany, 64(6), 1637-1647 (2013-02-05)
Gibberellic acid (GA) can regulate many plant developmental processes. GAST1 has been identified as a GA-stimulated transcript, and Arabidopsis GAST-like genes are known to constitute the GASA family. However, the functions of most GASA genes are not clear at present.
Pia Guadalupe Dominguez et al.
Plant physiology, 161(3), 1486-1500 (2013-01-11)
Asr (for ABA, stress, ripening) genes are exclusively found in the genomes of higher plants, and the encoded proteins have been found localized both to the nucleus and cytoplasm. However, before the mechanisms underlying the activity of ASR proteins can
Claus Schwechheimer et al.
Current opinion in plant biology, 12(1), 57-62 (2008-10-22)
Gibberellic acid (GA) promotes a range of developmental and growth processes in plants, the most well-known being germination, elongation growth and flowering time. DELLA repressors are the key players of the pathway. Their presence or their GA-dependent turnover via the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service